Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ova (329-337) | 276889-40-8 | >98.7% | 924.95 | 10 MG
SDP

Catalog No. 5000424286
Encompass_Preferred

OVA (329-337) is a 9-amino acid core epitope (329-337) found in the C-terminal end of the OVA peptide. It is intended for research use only and is not sold to patients. The product is shipped at room temperature within the continental US and should be stored sealed, away from moisture.

  • Stable for 2 years at -80°C in powder form
  • Stable for 1 year at -20°C in powder form
  • Stable for 6 months at -80°C if in solvent
  • Stable for 1 month at -20°C if in solvent
  • Recommended for research use only

Catalog No. 50-004-24286 Supplier Medchemexpress LLC Supplier No. HYP253110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

OVA (329-337) is a 9-amino acid core epitope (329-337) found in the C-terminal end of the OVA peptide. It is intended for research use only and is not sold to patients. The product is shipped at room temperature within the continental US and should be stored sealed, away from moisture.

  • Stable for 2 years at -80°C in powder form
  • Stable for 1 year at -20°C in powder form
  • Stable for 6 months at -80°C if in solvent
  • Stable for 1 month at -20°C if in solvent
  • Recommended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.