Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ova (329-337) | 276889-40-8 | 98.7% | 924.95 | 25 MG
SDP

Catalog No. 5000426700
Encompass_Preferred

OVA (329-337) is a 9-amino acid core epitope found at the C-terminal end of the OVA peptide. This peptide has demonstrated the ability to induce half-maximal proliferation in DO11.10 and OT-II cells at concentrations of 25 μM and 30 μM, respectively. It is typically a white to off-white solid and is soluble in H2O at 1.82 mg/mL (1.97 mM), though it is insoluble or slightly soluble in DMSO. This product is intended for research use only.

  • 9-amino acid core epitope from OVA peptide
  • Induces half-maximal proliferation in DO11.10 and OT-II cells
  • White to off-white solid appearance
  • Soluble in H2O at 1.82 mg/mL (1.97 mM)
  • For research use only
  • Recommended storage: sealed, away from moisture
  • Powder storage: -80°C for 2 years; -20°C for 1 year
  • In solvent storage: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-26700 Supplier Medchemexpress LLC Supplier No. 25MGHYP2531
May include imposed supplier surcharges.
Add to Cart
Add to Cart

OVA (329-337) is a 9-amino acid core epitope found at the C-terminal end of the OVA peptide. This peptide has demonstrated the ability to induce half-maximal proliferation in DO11.10 and OT-II cells at concentrations of 25 μM and 30 μM, respectively. It is typically a white to off-white solid and is soluble in H2O at 1.82 mg/mL (1.97 mM), though it is insoluble or slightly soluble in DMSO. This product is intended for research use only.

  • 9-amino acid core epitope from OVA peptide
  • Induces half-maximal proliferation in DO11.10 and OT-II cells
  • White to off-white solid appearance
  • Soluble in H2O at 1.82 mg/mL (1.97 mM)
  • For research use only
  • Recommended storage: sealed, away from moisture
  • Powder storage: -80°C for 2 years; -20°C for 1 year
  • In solvent storage: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.