Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OVA (55-62) | 151992-56-2 | 99.6% | 1004.30 | 10 MG
SDP

Catalog No. 5000424200
Encompass_Preferred

OVA 55-62 is a fragmented peptide of OVA (ovalbumin) antigen. This peptide is designed to bind to the mouse MHC class I molecule, H2-Kb, making it suitable for research applications in immunology.

  • Fragmented peptide of ovalbumin antigen
  • Binds to mouse MHC class I molecule, H2-Kb
  • High purity: 99.6%
  • Sequence: Lys-Val-Val-Arg-Phe-Asp-Lys-Leu
  • Appearance: White to off-white solid
  • Soluble in H2O
  • For research use only

Catalog No. 50-004-24200 Supplier Medchemexpress LLC Supplier No. HYP249410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OVA 55-62 is a fragmented peptide of OVA (ovalbumin) antigen. This peptide is designed to bind to the mouse MHC class I molecule, H2-Kb, making it suitable for research applications in immunology.

  • Fragmented peptide of ovalbumin antigen
  • Binds to mouse MHC class I molecule, H2-Kb
  • High purity: 99.6%
  • Sequence: Lys-Val-Val-Arg-Phe-Asp-Lys-Leu
  • Appearance: White to off-white solid
  • Soluble in H2O
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.