Learn More
BRINSEA PRODUCTS Ova-Easy 100 Cabinet Incubators Evaporating Blocks
Catalog No. NC2081705
Encompass_Preferred
These woven paper blocks are designed to increase humidity in the Brinsea OVA-Easy 100 Advance Series II cabinet incubators. The blocks locate in the water reservoir of the incubators and allow you to reach higher levels of humidity by increasing the evaporating surface area. These blocks are necessary when using the Advance humidity pump (part # NC1728213) to provide fully automatic control of humidity.
Catalog No. NC2081705 Supplier BRINSEA PRODUCTS Supplier No. 14.9310H
These woven paper blocks are designed to increase humidity in the Brinsea OVA-Easy 100 Advance Series II cabinet incubators. The blocks locate in the water reservoir of the incubators and allow you to reach higher levels of humidity by increasing the evaporating surface area. These blocks are necessary when using the Advance humidity pump (part # NC1728213) to provide fully automatic control of humidity.