Learn More
BRINSEA PRODUCTS Ova-Easy 580 Advance Series II Cabinet Incubator
Catalog No. NC1665284
Encompass_Preferred
Like all 4 models in the Brinsea cabinet incubator range, the Ova-Easy 580 Advance Series II is designed to offer the best possible performance and is fitted with Brinsea’s proven Advance fully digital control system which is both accurate and easy to use. Incubation temperature (in °C or °F) and humidity are continuously displayed along with temperature alarms and egg turning status. The unique laminar airflow, coupled with high levels of insulation mean that temperature is extremely evenly distributed throughout the egg chamber, ensuring excellent hatch rates time after time. It comes with many sophisticated features which include room temperature alarms and a single button press to automatically level the shelves for easy tray access. An external humidity pump can also be added to provide fully automatic control of humidity. Supplied with 12 universal egg trays with adjustable dividers to accommodate a wide range of egg sizes simultaneously.
Catalog No. NC1665284 Supplier BRINSEA PRODUCTS Supplier No. MJ5823C
Like all 4 models in the Brinsea cabinet incubator range, the Ova-Easy 580 Advance Series II is designed to offer the best possible performance and is fitted with Brinsea’s proven Advance fully digital control system which is both accurate and easy to use. Incubation temperature (in °C or °F) and humidity are continuously displayed along with temperature alarms and egg turning status. The unique laminar airflow, coupled with high levels of insulation mean that temperature is extremely evenly distributed throughout the egg chamber, ensuring excellent hatch rates time after time. It comes with many sophisticated features which include room temperature alarms and a single button press to automatically level the shelves for easy tray access. An external humidity pump can also be added to provide fully automatic control of humidity. Supplied with 12 universal egg trays with adjustable dividers to accommodate a wide range of egg sizes simultaneously.