Learn More
BRINSEA PRODUCTS Ova-Easy Advance Humidity Pump
Catalog No. NC1728213
Encompass_Preferred
The Ova-Easy Humidity Pump is specifically designed to work seamlessly with the Brinsea Ova-Easy Advance Series II cabinet incubators and hatchers. It includes a peristaltic pump and an external water reservoir, which eliminates the chances of water in the incubator or hatcher harboring bacteria. Easy to set up, this pump stands beside the incubator or hatcher and is connected to it by an electrical lead and a flexible silicone tube for water supply. Incubation humidity settings are controlled directly from the incubator's intuitive digital menu, and the achieved relative humidity (%RH) is continuously shown on the incubator’s digital display. The Brinsea Ova-Easy Advance cabinet incubators or hatchers, coupled with the humidity pump, effectively maintain the humidity level precisely at the level the user sets, irrespective of changes in room humidity.
Catalog No. NC1728213 Supplier BRINSEA PRODUCTS Supplier No. USAD25
The Ova-Easy Humidity Pump is specifically designed to work seamlessly with the Brinsea Ova-Easy Advance Series II cabinet incubators and hatchers. It includes a peristaltic pump and an external water reservoir, which eliminates the chances of water in the incubator or hatcher harboring bacteria. Easy to set up, this pump stands beside the incubator or hatcher and is connected to it by an electrical lead and a flexible silicone tube for water supply. Incubation humidity settings are controlled directly from the incubator's intuitive digital menu, and the achieved relative humidity (%RH) is continuously shown on the incubator’s digital display. The Brinsea Ova-Easy Advance cabinet incubators or hatchers, coupled with the humidity pump, effectively maintain the humidity level precisely at the level the user sets, irrespective of changes in room humidity.