Learn More
Medchemexpress LLC HY-P1771A 1mg Medchemexpress, OVA G4 peptide (TFA) CAS: Purity:>98%
SDP
Catalog No. 501934244
Encompass_Preferred
Medchemexpress, HY-P1771A 1mg OVA G4 peptide (TFA) CAS: OVA G4 peptide TFA is a variant of the agonist ovalbumin (OVA) peptide SIINFEKL (257-264). SIINFEKL is routinely used to stimulate ovalbumin-specific T cells and to test new vaccine adjuvants can form a stable hydrogel [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4244 Supplier Medchemexpress LLC Supplier No. HYP1771A1MG
Add to Cart
Add to Cart
Medchemexpress, HY-P1771A 1mg OVA G4 peptide (TFA) CAS: OVA G4 peptide TFA is a variant of the agonist ovalbumin (OVA) peptide SIINFEKL (257-264). SIINFEKL is routinely used to stimulate ovalbumin-specific T cells and to test new vaccine adjuvants can form a stable hydrogel [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.