Learn More
AnaSpec OVA (257-264) Peptide Fragment, Unconjugated, Lyophilized
Supplier:  AnaSpec AS601931
Encompass
OVA (257-264)[SIINFEKL] ; MW 963.2; (1 mg) This is a class I (Kb)-restricted peptide epitope of OVA, an octameric peptide from ovalbumin presented by the class I MHC molecule, H-2Kb.
Catalog No. 50-843-885