Learn More
CPC Scientific H-Ser-Ile-Ile-Asn-Phe-Glu-Lys-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MISC012B
Encompass
Ovalbumin-derived peptide known to arise from a natural processing of ovalbumin H-2b mouse cells. Subcutaneous immunization with this peptide could induce OVA-specific, CD8+CTLs that lyse the OVA-expressing target.SEQUENCE: H-Ser-Ile-Ile-Asn-Phe-Glu-Lys-Leu-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: SIINFEKLMOLECULAR FORMULA: C45H74N10O13MOLECULAR WEIGHT: 963.13STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [138831-86-4]SYNONYMS: OVA (257-264) (chicken), OVA peptide (257-264), Ovalbumin (257-264) (chicken)RESEARCH AREA: MiscellaneousREFERENCES:C. M. Celluzzi et al., J. Invest Dermatol.,108(5), 716 (1997).SKU(s): MISC-012A, MISC-012B, MISC-012C
Catalog No. 50-210-9736