Learn More
Medchemexpress LLC OVA Peptide(257-264) TFA | 1262751-08-5 | >99.9% | 1077.15 | 100 MG
SDP
Catalog No. 5000422206
Encompass_Preferred
OVA Peptide(257-264) TFA is a class I (Kb)-restricted peptide epitope of ovalbumin. This octameric peptide is presented by the class I MHC molecule, H-2Kb, and is primarily intended for research use.
  • Class I (Kb)-restricted peptide epitope
  • Derived from ovalbumin
  • Presented by the class I MHC molecule, H-2Kb
  • Intended for research use
Catalog No. 50-004-22206 Supplier Medchemexpress LLC Supplier No. HYP1489A100MG
OVA Peptide(257-264) TFA is a class I (Kb)-restricted peptide epitope of ovalbumin. This octameric peptide is presented by the class I MHC molecule, H-2Kb, and is primarily intended for research use.
  • Class I (Kb)-restricted peptide epitope
  • Derived from ovalbumin
  • Presented by the class I MHC molecule, H-2Kb
  • Intended for research use