Learn More
Medchemexpress LLC HY-P0286 1mg Medchemexpress, OVA Peptide (323-339) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02861MG
Encompass_Preferred
Medchemexpress, HY-P0286 1mg OVA Peptide (323-339) CAS: OVA Peptide (323-339) represents a T and B cell epitope of Ovalbumin (Ova), which is important in the generation and development of immediate hypersensitivity responses in BALB/c mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4097