Learn More
Genscript Corporation OVA Peptide (257-264)
Supplier:  Genscript Corporation RP10611
Encompass
This is a class I (Kb)-restricted peptide epitope of OVA, an octameric peptide from ovalbumin presented by the class I MHC molecule, H-2Kb.
Catalog No. NC1540274