Learn More
AnaSpec OVA-T4 Peptide, SIITFEKL, pT4, OVA (257-264) Variant, 1 mg
Supplier:  AnaSpec AS64403
Encompass
OVA-T4 Peptide, SIITFEKL, pT4, OVA (257-264) Variant[SIITFEKL] ; MW 950.2; (1 mg) T4 peptide (SIITFEKL) is a variant of the agonist ovalbumin (OVA) peptide (257-264), SIINFEKL. OVA Peptide is a class I (Kb)-restricted peptide epitope of ovalbumin presented by the class I MHC (major histocompatibility complex) molecule, H-2Kb (class I genes of the mouse MHC).
Catalog No. NC0783583