Learn More
Syndel USA Syndel Ovadine (PVP Iodine), 4 by 1 gal
SDP
Supplier:  Syndel USA OVADINE4/1GAL
Encompass
Syndels industry standard, non-corrosive buffered PVP Iodine solution provides 1% available iodine for the disinfection of fish eggs.
Catalog No. NC0224825