Learn More
Electron Microscopy Sciences Maxtaform Oval Hole Grid 2 x 1 mm hole, Cu/Rh
SDP
Catalog No. 5027582
Encompass_Preferred
High Grade Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness. Copper grids with one surface coated with inert Rhodium. This coating will eliminate tarnishing, give side identification, and reduce the bar thickness.
Size: 2 x 1 mm
Material: Cu/Rh
Catalog No. 50-275-82 Supplier Electron Microscopy Sciences Supplier No. M2010CR
High Grade Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness. Copper grids with one surface coated with inert Rhodium. This coating will eliminate tarnishing, give side identification, and reduce the bar thickness.
Size: 2 x 1 mm
Material: Cu/Rh