Learn More
Electron Microscopy Sciences Maxtaform Oval Hole Grid 2 x 1 mm hole, Nickel
SDP
Catalog No. 5027581
Encompass_Preferred
High Grade Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Size: 2 x 1 mm
Material: Nickel
Catalog No. 50-275-81 Supplier Electron Microscopy Sciences Supplier No. M2010NI
High Grade Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Size: 2 x 1 mm
Material: Nickel