Learn More
Electron Microscopy Sciences Embra Grid with 0.4x2 mm slot, Aluminum
SDP
Catalog No. 5027585
Encompass_Preferred
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 50 micron thick for oval hole grids.
I.D.: 0.4x2 mm
Material: Aluminum
Catalog No. 50-275-85 Supplier Electron Microscopy Sciences Supplier No. E0420AL
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 50 micron thick for oval hole grids.
I.D.: 0.4x2 mm
Material: Aluminum