Learn More
Electron Microscopy Sciences Embra Grid with 0.4x2 mm slot, Stainless
SDP
Catalog No. 5027584
Encompass_Preferred
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 50 micron thick for oval hole grids.
I.D.: 0.4x2 mm
Material: Stainless
Catalog No. 50-275-84 Supplier Electron Microscopy Sciences Supplier No. E0420SS
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 50 micron thick for oval hole grids.
I.D.: 0.4x2 mm
Material: Stainless