Learn More
Electron Microscopy Sciences Embra Grid with 0.4x2 mm slot, Titanium
SDP
Catalog No. 5027583
Encompass_Preferred
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 50 micron thick for oval hole grids.
I.D.: 0.4x2 mm
Material: Titanium
Catalog No. 50-275-83 Supplier Electron Microscopy Sciences Supplier No. E0420TI
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 50 micron thick for oval hole grids.
I.D.: 0.4x2 mm
Material: Titanium