Learn More
Sigma Aldrich Fine Chemicals Biosciences Rare earth extra power stir bars oval, diam. 5 mm, L 10 mm | 3EA
Catalog No. NC2025725
Encompass
Rare earth extra power stir bars oval, diam. 5 mm, L 10 mm | 3EA
Catalog No. NC2025725 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6716223EA
Rare earth extra power stir bars oval, diam. 5 mm, L 10 mm | 3EA