Learn More
Medchemexpress LLC Ovalbumin (154-159) | 1370698-94-4 | 99.9% | 672.77 | 10 MG
SDP
Catalog No. 5000397658
Encompass_Preferred
Ovalbumin (154-159) is a protein fragment composed of residues 154-159 of chicken ovalbumin. It is also identified as a poorly conserved sequence (Thr-Asn-Gly-Ile-Ile-Arg) within the α-subunit of the Torpedo acetylcholine receptor, specifically located in the α150-164 epitope region. This fragment demonstrates significant angiotensin-converting enzyme (ACE) inhibitory activity.
  • Suitable for use as a standard protein sample for molecular weight estimation in small-angle X-ray scattering experiments.
  • Commonly utilized as a molecular weight standard in SDS-PAGE.
  • Used for research related to hypertension.
Catalog No. 50-003-97658 Supplier Medchemexpress LLC Supplier No. HYP427910MG
Ovalbumin (154-159) is a protein fragment composed of residues 154-159 of chicken ovalbumin. It is also identified as a poorly conserved sequence (Thr-Asn-Gly-Ile-Ile-Arg) within the α-subunit of the Torpedo acetylcholine receptor, specifically located in the α150-164 epitope region. This fragment demonstrates significant angiotensin-converting enzyme (ACE) inhibitory activity.
  • Suitable for use as a standard protein sample for molecular weight estimation in small-angle X-ray scattering experiments.
  • Commonly utilized as a molecular weight standard in SDS-PAGE.
  • Used for research related to hypertension.