Learn More
Medchemexpress LLC Ovalbumin (154-159) | 1370698-94-4 | 99.94% | 672.77 | 5 MG
SDP
Catalog No. 5000397617
Encompass_Preferred
Ovalbumin (154-159) is a protein fragment consisting of residues 154-159 of chicken ovalbumin. It exhibits significant angiotensin-converting enzyme (ACE) inhibitory activity and can serve as a standard protein sample for molecular weight estimation in small-angle X-ray scattering experiments. It is also commonly used as a molecular weight standard in SDS-PAGE and for research on hypertension.
  • Protein fragment from chicken ovalbumin
  • Exhibits significant ACE inhibitory activity
  • Used as a standard protein sample for molecular weight estimation
  • Commonly used as a molecular weight standard in SDS-PAGE
  • Applied in research on hypertension
Catalog No. 50-003-97617 Supplier Medchemexpress LLC Supplier No. HYP42795MG
Ovalbumin (154-159) is a protein fragment consisting of residues 154-159 of chicken ovalbumin. It exhibits significant angiotensin-converting enzyme (ACE) inhibitory activity and can serve as a standard protein sample for molecular weight estimation in small-angle X-ray scattering experiments. It is also commonly used as a molecular weight standard in SDS-PAGE and for research on hypertension.
  • Protein fragment from chicken ovalbumin
  • Exhibits significant ACE inhibitory activity
  • Used as a standard protein sample for molecular weight estimation
  • Commonly used as a molecular weight standard in SDS-PAGE
  • Applied in research on hypertension