Learn More
Bachem Ovalbumin (257-264) (chicken) Trifluoroacetate
SDP
Supplier:  Bachem 4033142.0005
Encompass_Preferred
Ovalbumin (257-264) (chicken) Trifluoroacetate, H-Ser-Ile-Ile-Asn-Phe-Glu-Lys-Leu-OH, 138831-86-4, C45H74N10O13, Mr 963.14, 5mg. OVA (257-264) (chicken), OVA peptide (257-264). SIINFEKL, an ovalbumin-derived peptide, is known to arise from natural processing of ovalbumin H-2b mouse cells. Cytotoxic T lymphocytes (CTL) from the ovalbumin-hsp70-immunized mice and a highly effective CTL clone (4G3) raised against ovalbumin-expressing EL4 tumor cells (EG7-OVA) were equally effective in terms of the concentration of this compound required for half-maximal lysis in a CTL assay.
Catalog No. 50-259-886