Learn More
Apexbio Technology LLC ovalbumin (324-338) [Gallus gallus]/[Coturnix coturnix], 25mg
SDP
Supplier:  Apexbio Technology LLC A108225
Encompass_Preferred
Ovalbumin (abbreviated OVA) is the main protein found in egg white, making up 60-65% of the total protein(1).Ovalbumin is an important protein in several different areas of research, including:general studies of protein structure and properties (because it is available in large quantities). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4685