Learn More
Bioworld Ovalbumin, 1 g
Catalog No. 50253909
Encompass_Preferred
Albumin hen egg whiteCAS: 9006-59-1Purity: >97%MW 44287 Da Storage: 2-8°C
Catalog No. 50-253-909 Supplier Bioworld Supplier No. 220700131
Albumin hen egg whiteCAS: 9006-59-1Purity: >97%MW 44287 Da Storage: 2-8°C