Learn More
Alkali Scientific Ovalbumin [Albumin from Chicken Egg White], 25 G
SDP
Catalog No. 5000661564
Encompass_Preferred
Ovalbumin (Albumin from Chicken Egg White) is a highly purified protein that plays a key role in many laboratory applications. This 25g pack is ideal for researchers working with large-scale experiments, including immunological studies, protein assays, and cell culture protocols. Ovalbumin is commonly used as a carrier protein in immunization procedures and as a negative control in protein assays. Its high purity and availability make it essential for investigations into protein function, antigen-antibody interactions, and enzyme inhibition. This reagent is particularly useful in preparing standard protein solutions for quantification assays, enzyme-linked immunosorbent assays (ELISA), and other immunological assays.
Catalog No. 50-006-61564 Supplier Alkali Scientific Supplier No. OVA002CP
Ovalbumin (Albumin from Chicken Egg White) is a highly purified protein that plays a key role in many laboratory applications. This 25g pack is ideal for researchers working with large-scale experiments, including immunological studies, protein assays, and cell culture protocols. Ovalbumin is commonly used as a carrier protein in immunization procedures and as a negative control in protein assays. Its high purity and availability make it essential for investigations into protein function, antigen-antibody interactions, and enzyme inhibition. This reagent is particularly useful in preparing standard protein solutions for quantification assays, enzyme-linked immunosorbent assays (ELISA), and other immunological assays.