Learn More
Santa Cruz Biotechnology Ovalbumin Antibody (3G2E1D9) Alexa Fluor® 647, 200 µg/ml
Catalog No. NC3938994
Encompass
Ovalbumin Antibody (3G2E1D9) is a mouse monoclonal IgG1 κ Ovalbumin antibody, cited in 6 publications, provided at 200 µg/mlraised against ovalbumin of Chicken originOvalbumin Antibody (3G2E1D9) is recommended for detection of Ovalbumin of avian origin by WB and IPAnti-Ovalbumin Antibody (3G2E1D9) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM
Catalog No. NC3938994 Supplier Santa Cruz Biotechnology Supplier No. SC65984 AF647
Ovalbumin Antibody (3G2E1D9) is a mouse monoclonal IgG1 κ Ovalbumin antibody, cited in 6 publications, provided at 200 µg/mlraised against ovalbumin of Chicken originOvalbumin Antibody (3G2E1D9) is recommended for detection of Ovalbumin of avian origin by WB and IPAnti-Ovalbumin Antibody (3G2E1D9) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM