Learn More
Medchemexpress LLC Ovalbumin-Cy5 | 45 kDa | 1 MG
SDP
Catalog No. 5000426542
Encompass_Preferred
Ovalbumin-Cy5 is a fluorescent conjugate of Ovalbumin, a well-characterized protein, with the cyanine dye Cy5. Cy5 is a far-red fluorescent dye commonly used in various biological applications due to its high extinction coefficient and fluorescence quantum yield. This conjugate can be utilized as a tracer in imaging, flow cytometry, and other assays requiring a fluorescently labeled protein.
  • Fluorescent conjugate of Ovalbumin
  • Provides far-red fluorescence
  • Suitable for imaging and flow cytometry
  • Used as a tracer in various biological assays
Catalog No. 50-004-26542 Supplier Medchemexpress LLC Supplier No. HYNP0551MG
Ovalbumin-Cy5 is a fluorescent conjugate of Ovalbumin, a well-characterized protein, with the cyanine dye Cy5. Cy5 is a far-red fluorescent dye commonly used in various biological applications due to its high extinction coefficient and fluorescence quantum yield. This conjugate can be utilized as a tracer in imaging, flow cytometry, and other assays requiring a fluorescently labeled protein.
  • Fluorescent conjugate of Ovalbumin
  • Provides far-red fluorescence
  • Suitable for imaging and flow cytometry
  • Used as a tracer in various biological assays