Learn More
Worthington Biochemical Corporation Ovalbumin, Low Endo™, Purified, 100mg
SDP
Catalog No. NC0516480
Encompass
Ovalbumin, purified to remove endotoxin and tested. A dialyzed, lyophilized powder containing ≤ 1 EU/mg. Store at 2-8°C. Ovalbumin is a glycoprotein with molecular weight of 45 kDa. The molecule consists of a polypeptide with up to two phosphate groups per mole and a side chain of mannose and glucosamine residues.
Catalog No. NC0516480 Supplier Worthington Biochemical Corporation Supplier No. LS003061
Ovalbumin, purified to remove endotoxin and tested. A dialyzed, lyophilized powder containing ≤ 1 EU/mg. Store at 2-8°C. Ovalbumin is a glycoprotein with molecular weight of 45 kDa. The molecule consists of a polypeptide with up to two phosphate groups per mole and a side chain of mannose and glucosamine residues.