Learn More
Biomatik Corporation Ovalbumin (Ova) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU29966
Encompass_Preferred
Ovalbumin (Ova) Polyclonal Antibody is a Universal Polyclonal Antibody to Ovalbumin (OVA). The Ovalbumin (Ova) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Universal. Applications: WB, IHC, ICC, IP. Target Synonyms: Ovalbumin (OVA). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6627