Learn More
Worthington Biochemical Corporation OVALBUMIN PURIFIED 100MG
SDP
Supplier:  Worthington Biochemical Corporation LS003056
Encompass
Chromatographically purified. Major protein of egg white, with a molecular weight of 45 kDa. A dialyzed, lyophilized powder.
Catalog No. NC9639031