Learn More
Cusabio Technology LLC ovalbumin specific IgE
Catalog No. 5000049013
Encompass_Preferred
This ELISA kit detects mouse ovalbumin-specific IgE (OVA sIgE) in serum and plasma samples offering qualitative analysis through an indirect immunoassay method The assay can be completed within 1-5 hours and requires a sample volume of 50-100 l Detection is performed at 450 nm ensuring accurate measurement of OVA sIgE levels Designed for research applications this kit is ideal for studying allergic sensitization and immune responses to ovalbumin in murine models providing reliable results for evaluating IgE-mediated hypersensitivity Its standardized protocol ensures consistency making it suitable for laboratories focused on immunology or allergy research
Catalog No. 50-000-49013 Supplier Cusabio Technology LLC Supplier No. CSBE08914M
This ELISA kit detects mouse ovalbumin-specific IgE (OVA sIgE) in serum and plasma samples offering qualitative analysis through an indirect immunoassay method The assay can be completed within 1-5 hours and requires a sample volume of 50-100 l Detection is performed at 450 nm ensuring accurate measurement of OVA sIgE levels Designed for research applications this kit is ideal for studying allergic sensitization and immune responses to ovalbumin in murine models providing reliable results for evaluating IgE-mediated hypersensitivity Its standardized protocol ensures consistency making it suitable for laboratories focused on immunology or allergy research