Learn More
Cusabio Technology LLC ovalbumin specific IgG
Catalog No. 5000050288
Encompass_Preferred
This ELISA kit detects OVA sIgG (Rat ovalbumin specific IgG) in serum and plasma samples using a qualitative indirect assay format Designed for specificity and reliability the kit employs an indirect detection method to identify the presence of OVA-specific IgG antibodies providing results within 1 to 5 hours Each test requires 50-100 L of sample making it suitable for studies with limited sample availability The assay utilizes a 450 nm wavelength for optical density measurements ensuring standardized readouts Ideal for immunological research this kit enables efficient evaluation of immune responses to ovalbumin in rat models supporting applications in allergy studies vaccine development and autoimmune research Its optimized protocol ensures consistent performance across experimental conditions
Catalog No. 50-000-50288 Supplier Cusabio Technology LLC Supplier No. CSBE13486R
This ELISA kit detects OVA sIgG (Rat ovalbumin specific IgG) in serum and plasma samples using a qualitative indirect assay format Designed for specificity and reliability the kit employs an indirect detection method to identify the presence of OVA-specific IgG antibodies providing results within 1 to 5 hours Each test requires 50-100 L of sample making it suitable for studies with limited sample availability The assay utilizes a 450 nm wavelength for optical density measurements ensuring standardized readouts Ideal for immunological research this kit enables efficient evaluation of immune responses to ovalbumin in rat models supporting applications in allergy studies vaccine development and autoimmune research Its optimized protocol ensures consistent performance across experimental conditions