Learn More
Cusabio Technology LLC ovalbumin specific IgG
Catalog No. 5000048349
Encompass_Preferred
This ELISA kit detects OVA sIgG (Guinea pig ovalbumin specific IgG) in serum and plasma samples using an indirect qualitative method The assay requires 50-100 l of sample and delivers results within 1-5 hours Absorbance is measured at 450 nm to determine the presence of OVA sIgG antibodies making it suitable for applications requiring specific and efficient detection of immune responses The kit is optimized for ease of use and reliability providing researchers with a robust tool for studying immunological profiles in guinea pig models or related diagnostic evaluations
Catalog No. 50-000-48349 Supplier Cusabio Technology LLC Supplier No. CSBE10139GU
This ELISA kit detects OVA sIgG (Guinea pig ovalbumin specific IgG) in serum and plasma samples using an indirect qualitative method The assay requires 50-100 l of sample and delivers results within 1-5 hours Absorbance is measured at 450 nm to determine the presence of OVA sIgG antibodies making it suitable for applications requiring specific and efficient detection of immune responses The kit is optimized for ease of use and reliability providing researchers with a robust tool for studying immunological profiles in guinea pig models or related diagnostic evaluations