Learn More
Bioworld Ovalbumin Standard, 10 mL
Catalog No. 50253912
Encompass_Preferred
Ovalbumin, Protein Standard2mg/mlSold as 5 x 2mL.Supplied in 0.8% NaCl with 10 mM Sodium Azide
Catalog No. 50-253-912 Supplier Bioworld Supplier No. 220700141
Ovalbumin, Protein Standard2mg/mlSold as 5 x 2mL.Supplied in 0.8% NaCl with 10 mM Sodium Azide