Learn More
Discovery Life Sciences Ovarian Cancer Tx Naive PBMC
SDP
Catalog No. 5000804109
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04109 Supplier Discovery Life Sciences Supplier No. 120290
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays