Learn More
VistaLab™ Technologies Ovation™ Adjustable-Volume Pipetter Starter Set PROMO
SDP
Catalog No. 22146685TI Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
2-200μL, 20-200μL, 100-1000μL
Catalog No. Volume (Metric)
22-146-685TI 2-200μL, 20-200μL, 100-1000μL
1 options
Catalog No. 22-146-685TI Supplier VistaLab™ Technologies Supplier No. 22146685TI
Add to Cart
Add to Cart
Ovation Quick-Set Volume Started Pack allows five of the laboratory's most frequently used volumes to be set up and stored for routine use and later recall.
  • Other volume settings can be selected at precise increments throughout the pipetter's range
  • A low-force, form-fitting plunger manually controls aspiration and dispensing speeds with minimal effort
Specifications
Product Line Ovation
Type Adjustable Volume Pipette Starter Set
Volume (Metric) 2-200μL, 20-200μL, 100-1000μL
Includes One 2 to 20 μL, one 20 to 200 μL, and one 100 to 1000 μL pipettes, 3 Ovation Shelf Hangers, 3 racks of Ovation tips