Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Replacement Duracell™ Ultra CR2 Battery for Ovation™ QS Pipette
SDP

Catalog No. 22146667 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Battery Type:
CR2032
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Type
22-146-667 CR2032
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-146-667 Supplier VistaLab™ Technologies Supplier No. 90574001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3 volt lithium battery with an expected life of one year or more of typical use.

Specifications

Product Type CR2 Battery
Battery Type CR2032
For Use With (Equipment) Ovation Adjustable Volume Pipettes
Quantity 1 Ea.
Length (English) 5 in.
Width (English) 3 in.
Height (English) 1 in.
Voltage 3 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.