Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Ovation M Hook and Top Cover Replacement Kit
SDP

Catalog No. 22147640 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Ovation M 0.2 - 2 μL
Ovation M 1 - 10 μL
Ovation M 10 - 100 μL
Ovation M 100 - 1000 μL
Ovation M 2 - 20 μL
Ovation M 20 - 200 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-147-640 Ovation M 100 - 1000 μL
22-147-635 Ovation M 0.2 - 2 μL
22-147-636 Ovation M 1 - 10 μL
22-147-637 Ovation M 2 - 20 μL
22-147-638 Ovation M 10 - 100 μL
22-147-639 Ovation M 20 - 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-147-640 Supplier VistaLab™ Technologies Supplier No. 90704013
Only null left
Add to Cart
Add to Cart

Maintenance part

Hook and Top Cover Replacement for Ovation M Pipettes

Specifications

Description Replacement kit for hook and top cover of Ovation M Pipettes
For Use With Pipettes
Material Plastic
Sterility Non-sterile
Volume (Metric) 100 to 1000 μL
For Use With (Equipment) Ovation M 100 - 1000 μL
Quantity 1 Ea.
Length (English) 5 in.
Width (English) 3 in.
Height (English) 1 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.