Learn More
VistaLab™ Technologies Ovation Nozzle Seal E8 & E12
SDP
Catalog No. 22147652 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
2-125 μL, 5-250 μL
Catalog No. Volume (Metric)
22-147-652 2-125 μL, 5-250 μL
1 options
Catalog No. 22-147-652 Supplier VistaLab™ Technologies Supplier No. 90602012
Add to Cart
Add to Cart
Maintenance part
Nozzle seals for 2-125 μL and 5-250 μL E8 & E12 Ovation pipettes
Specifications
Description Medium Nozzle Seals
For Use With Pipettes
Material Plastic
Sterility Non-sterile
Volume (Metric) 2-125 μL, 5-250 μL
For Use With (Equipment) E8 & E12 Ovation Pipettes
Quantity 24 seals/bag
Length (English) 2 in.
Width (English) 2 in.
Height (English) 5 in.