Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Ovation™ Pipet Accessories: Reconditioning Nozzle Kit
SDP

Catalog No. 22146245 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
For Use With (Equipment):
0.5 to 20 μL Ovation™ BioNatural™ Multichannel Pipette
2 to 125 μL, 5 to 250 μL Ovation™ BioNatural™ Multichannel Pipette
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-146-245 0.5 to 20 μL Ovation™ BioNatural™ Multichannel Pipette
22-146-249 2 to 125 μL, 5 to 250 μL Ovation™ BioNatural™ Multichannel Pipette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-146-245 Supplier VistaLab™ Technologies Supplier No. 90601011
Add to Cart
Add to Cart

For Ovation E8 & E12 Multichannel Pipets

  • Contains 2 nozzles, 24 nozzle seals, 24 filters & nozzle removal tool

Specifications

Description Small Nozzle Reconditioning Kit
For Use With Pipettes
Material Plastic
Sterility Non-sterile
Volume (Metric) 0.5 to 20 μL
For Use With (Equipment) 0.5 to 20 μL Ovation™ BioNatural™ Multichannel Pipette
Includes 2 Nozzles, 24 seals, 24 filters and tool
Quantity 1 Ea.
Length (English) 2 in.
Width (English) 2 in.
Height (English) 5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.