Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Ovation™ Pipet Accessories: Reconditioning Nozzle Kit
GSA_VASDP

Catalog No. 22146249 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
For Use With (Equipment):
0.5 to 20 μL Ovation™ BioNatural™ Multichannel Pipette
2 to 125 μL, 5 to 250 μL Ovation™ BioNatural™ Multichannel Pipette
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-146-249 2 to 125 μL, 5 to 250 μL Ovation™ BioNatural™ Multichannel Pipette
22-146-245 0.5 to 20 μL Ovation™ BioNatural™ Multichannel Pipette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-146-249 Supplier VistaLab™ Technologies Supplier No. 90602011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Ovation E8 & E12 Multichannel Pipets

  • Contains 2 nozzles, 24 nozzle seals, 24 filters & nozzle removal tool

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Description Medium Nozzle Reconditioning Kit
For Use With Pipettes
Material Plastic
Sterility Non-sterile
Volume (Metric) 2 to 125 μL, 5 to 250 μL
For Use With (Equipment) 2 to 125 μL, 5 to 250 μL Ovation™ BioNatural™ Multichannel Pipette
Includes 2 Nozzles, 24 seals, 24 filters and tool
Quantity 1 Ea.
Length (English) 2 in.
Width (English) 2 in.
Height (English) 5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.