Learn More
Aviva Systems Biology Corp Ovca2 Antibody - middle region (ARP50757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ovca2. It was validated on Western Blot. . Alternate names: RGD1564623ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-537