Learn More
Sheldon Manufacturing Inc OVEN, FORCED AIR, 5 CUFT, 115V
SDP
Catalog No. NC1484587
Encompass_Preferred
The SMO5 is equipped with a stainless steel interior for long life operation and easy cleaning.With triple wall construction, this oven’s walls minimize external wall temperature which meets CE requirements to keep work-spaces cool.These ovens are recommended for critical and analytical use in laboratory and pilot plant product research. Typical applications for the oven include aging, asphalt testing, curing, drying, pre-heating, conditioning and life cycle testing. This unit has a quick ramp-up time to 306°C for faster cycle times.This unit includes an advanced PID temperature control system for sensitive response.The SMO5’s forced air ovens deliver precise uniformity, air distribution and the peace of mind expected of the SHEL LAB brand.
Catalog No. NC1484587 Supplier Sheldon Manufacturing Inc Supplier No. SMO5
Add to Cart
Add to Cart
Request a Quote
The SMO5 is equipped with a stainless steel interior for long life operation and easy cleaning.With triple wall construction, this oven’s walls minimize external wall temperature which meets CE requirements to keep work-spaces cool.These ovens are recommended for critical and analytical use in laboratory and pilot plant product research. Typical applications for the oven include aging, asphalt testing, curing, drying, pre-heating, conditioning and life cycle testing. This unit has a quick ramp-up time to 306°C for faster cycle times.This unit includes an advanced PID temperature control system for sensitive response.The SMO5’s forced air ovens deliver precise uniformity, air distribution and the peace of mind expected of the SHEL LAB brand.