Learn More
Sheldon Manufacturing Inc OVEN FORCED AIR LRG REP-FX28-2
SDP
Catalog No. NC0734853
Encompass_Preferred
OVEN, FORCED AIR, 27.5 CU FT, DIGITAL WITH TIMER, 230V The SHEL LAB Model SMO28-2 is the largest general purpose oven on the market (also consider the SMO34HP-2 or SMO38HP-2 high performance ovens for the best uniformity and functionality). The independent overtemperature safety feature is user adjustable and provides added safety. The turbo blower-assisted forced airflow design coupled with a heavy-duty blower motor offers excellent temperature uniformity and fast recovery times. Airflow is directed evenly across all shelves and samples to ensure consistent drying, curing and baking results. These units are especially ideal for glassware drying in core facilities. More information available at www.shellab.com
Catalog No. NC0734853 Supplier Sheldon Manufacturing Inc Supplier No. SMO282
Add to Cart
Add to Cart
Request a Quote
OVEN, FORCED AIR, 27.5 CU FT, DIGITAL WITH TIMER, 230V The SHEL LAB Model SMO28-2 is the largest general purpose oven on the market (also consider the SMO34HP-2 or SMO38HP-2 high performance ovens for the best uniformity and functionality). The independent overtemperature safety feature is user adjustable and provides added safety. The turbo blower-assisted forced airflow design coupled with a heavy-duty blower motor offers excellent temperature uniformity and fast recovery times. Airflow is directed evenly across all shelves and samples to ensure consistent drying, curing and baking results. These units are especially ideal for glassware drying in core facilities. More information available at www.shellab.com