Learn More
MSC Betco Oven Fryer Grill Cleaner, Liquid, 1 Gallon, Nonfuming
Catalog No. 502836229
Encompass
This powerful oven, fryer, and grill cleaner is designed to effectively remove stubborn baked-on carbon and soil. Its unique formulation penetrates and breaks the chemical bond of deposits, allowing for fast and easy removal, unlike many foaming products that merely liquefy soils. It is nonfuming and nonflammable, making it a safer and more pleasant option for cleaning. This versatile cleaner can be used on various kitchen equipment, leaving behind a fresh fragrance.
  • Penetrates and removes baked-on deposits.
  • Nonfuming and nonflammable for safer use.
  • Breaks chemical bonds for easy soil removal.
  • Can be used on ovens, grills, and range hoods.
  • Leaves a fresh fragrance.
  • Supplied in a 1 gallon bottle for ample supply.
Catalog No. 50-283-6229 Supplier MSC Supplier No. 72529159
Add to Cart
Add to Cart
This powerful oven, fryer, and grill cleaner is designed to effectively remove stubborn baked-on carbon and soil. Its unique formulation penetrates and breaks the chemical bond of deposits, allowing for fast and easy removal, unlike many foaming products that merely liquefy soils. It is nonfuming and nonflammable, making it a safer and more pleasant option for cleaning. This versatile cleaner can be used on various kitchen equipment, leaving behind a fresh fragrance.
  • Penetrates and removes baked-on deposits.
  • Nonfuming and nonflammable for safer use.
  • Breaks chemical bonds for easy soil removal.
  • Can be used on ovens, grills, and range hoods.
  • Leaves a fresh fragrance.
  • Supplied in a 1 gallon bottle for ample supply.