Learn More
Electron Microscopy Sciences Oven Incubator 120 V
SDP
Catalog No. 5027591
Encompass_Preferred
This Oven/Incubator with an aluminum inner chamber, and venting will provide many years of long service in busy labs. The cast aluminum chamber provides for dependable temperature uniformity and the vent efficiently expels moisture and vapors. The unit has a magnetic latch that assures positive door closure and a thermometer port at the top of the oven that accommodates any partial immersion thermometer. The unit heats up rapidly (56 degree C in 9 minutes) and comes with a removable shelf.
Catalog No. 50-275-91 Supplier Electron Microscopy Sciences Supplier No. 6323010
This Oven/Incubator with an aluminum inner chamber, and venting will provide many years of long service in busy labs. The cast aluminum chamber provides for dependable temperature uniformity and the vent efficiently expels moisture and vapors. The unit has a magnetic latch that assures positive door closure and a thermometer port at the top of the oven that accommodates any partial immersion thermometer. The unit heats up rapidly (56 degree C in 9 minutes) and comes with a removable shelf.