Learn More
MSC Betco Oven Jell Cleaner, Gel, Lemon, Nonflammable, 1 Gallon
Catalog No. 502839132
Encompass
This oven and grill cleaner is formulated to cling to vertical surfaces, providing extended contact time for effective cleaning. Its jelling action quickly penetrates and dissolves tough baked-on and burnt-on deposits, making removal fast and easy. It is nonfuming and nonflammable, suitable for various food processing and service environments.
  • NSF International certified for use in all regulated food processing and food service environments.
  • Jelling action clings to surfaces to quickly penetrate and remove baked and burnt-on deposits.
  • Nonfuming and nonflammable.
  • Quickly dissolves even the toughest soils.
  • Breaks the chemical bond between the soil and oven surface, releasing baked on deposits.
Catalog No. 50-283-9132 Supplier MSC Supplier No. 73004095
This oven and grill cleaner is formulated to cling to vertical surfaces, providing extended contact time for effective cleaning. Its jelling action quickly penetrates and dissolves tough baked-on and burnt-on deposits, making removal fast and easy. It is nonfuming and nonflammable, suitable for various food processing and service environments.
  • NSF International certified for use in all regulated food processing and food service environments.
  • Jelling action clings to surfaces to quickly penetrate and remove baked and burnt-on deposits.
  • Nonfuming and nonflammable.
  • Quickly dissolves even the toughest soils.
  • Breaks the chemical bond between the soil and oven surface, releasing baked on deposits.