Learn More
MSC Betco Oven Jell Cleaner, Gel, 32 oz Bottle, NSF Certified
Catalog No. 502837493
Encompass
This fast-acting gel cleaner is formulated for ovens, grills, and range hoods. Its unique jelling action allows it to cling to vertical surfaces, providing prolonged contact time to dissolve tough, baked-on soils more effectively than foaming products. It breaks the chemical bond between soil and the oven surface for easy removal of deposits.
  • NSF International certified for use in food processing and service environments.
  • Jelling action clings to surfaces to penetrate and remove baked and burnt-on deposits.
  • Nonfuming and nonflammable.
  • Features a refreshing lemon fragrance.
  • Has a pH range of 12.5-14.0.
  • Contains sodium hydroxide for effective cleaning.
Catalog No. 50-283-7493 Supplier MSC Supplier No. 72969017
This fast-acting gel cleaner is formulated for ovens, grills, and range hoods. Its unique jelling action allows it to cling to vertical surfaces, providing prolonged contact time to dissolve tough, baked-on soils more effectively than foaming products. It breaks the chemical bond between soil and the oven surface for easy removal of deposits.
  • NSF International certified for use in food processing and service environments.
  • Jelling action clings to surfaces to penetrate and remove baked and burnt-on deposits.
  • Nonfuming and nonflammable.
  • Features a refreshing lemon fragrance.
  • Has a pH range of 12.5-14.0.
  • Contains sodium hydroxide for effective cleaning.