Learn More
RDP CORPORATION OVEN MODEL LCC1-51-5
Catalog No. NC3705245
Encompass
Oven model LCC1-51-5 (Standard Temperature Package) [Air Atmosphere, 260°C (500°F), 1.5°C/Minute HEPA Filter]HEPA (High-efficiency particulate air) FilterRecirculation air is 100% filtered through a 99.99% HEPA filter for class 100 or better operation.
Catalog No. NC3705245 Supplier RDP CORPORATION Supplier No. 01.024 LCC1515
Add to Cart
Add to Cart
Request a Quote
Oven model LCC1-51-5 (Standard Temperature Package) [Air Atmosphere, 260°C (500°F), 1.5°C/Minute HEPA Filter]HEPA (High-efficiency particulate air) FilterRecirculation air is 100% filtered through a 99.99% HEPA filter for class 100 or better operation.